- Description:Mouse Insulin like Growth Factor 1 ELISA kit (IGF1)
- Tests per kit:96 tests
Mouse Insulin like Growth Factor 1 ELISA kit (IGF1) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Insulin like Growth Factor 1 (IGF1) in Mouse in Plasma, Cell culture supernatant, Serum samples.