Mouse Cathepsin D ELISA Kit
Catalog # ABCAAB213845-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Cathepsin D ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse Cathepsin D ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Cathepsin D in Mouse in Cell Lysate, Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum samples.