Rat Kallikrein 10/K10 Antibody Pair - BSA and Azide Free
Catalog # ABCAAB253852-10X96
Supplier: Abcam
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:Kallikrein 10/K10
- Description:Rat Kallikrein 10/K10 antibody pair - BSA and azide free
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Rat Kallikrein 10/K10 antibody pair - BSA and Azide free.