ContactUsLinkComponent
Mouse Caspase-1 Antibody Pair - BSA and Azide free
Mouse Caspase-1 Antibody Pair - BSA and Azide free
Catalog # ABCAAB322485-10X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Mouse Caspase-1 Antibody Pair - BSA and Azide free
Catalog # ABCAAB322485-10X96
Supplier:  Abcam
New Product
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Mouse Caspase-1 Antibody Pair - BSA and Azide free
  • Tests per kit:
    10×96 tests
Specifications
About this item
Mouse Caspase-1 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse Caspase-1.