Mouse Eotaxin-2 ELISA Kit (CCL24)
Catalog # ABCAAB100681-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Eotaxin-2 ELISA kit (CCL24)
- Tests per kit:96 tests
Specifications
About this item
Mouse Eotaxin-2 ELISA Kit (CCL24) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Eotaxin-2 (CCL24) in Mouse in Plasma, Cell culture supernatant, Serum samples.