- Description:Rat MIP-1 beta ELISA kit (CCL4)
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Rat MIP-1 beta ELISA kit (CCL4) is a Sandwich (quantitative) ELISA kit for the measurement of Rat MIP-1 beta (CCL4) in Rat in cell culture supernatant, citrate plasma, EDTA plasma, heparin plasma, serum samples.