ContactUsLinkComponent
Human Syntaxin 1B ELISA Kit
Catalog # ABCAAB316901-96TST
Supplier:  Abcam
New Product
CAS Number:  
undefined
Human Syntaxin 1B ELISA Kit
Catalog # ABCAAB316901-96TST
Supplier:  Abcam
New Product
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Human Syntaxin 1B ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Human Syntaxin 1B ELISA kit is a single-wash 90-min Simplestep used to quantify Human Syntaxin 1B with a sensitivity of 16,949 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader.