Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-RUNX2 Rabbit Monoclonal Antibody [clone: EPR22858-106] (Alexa Fluor® 555)
Anti-RUNX2 Rabbit Monoclonal Antibody [clone: EPR22858-106] (Alexa Fluor® 555)
Catalog # ABCAAB318828-100UL
Supplier:  Abcam
New Product
Anti-RUNX2 Rabbit Monoclonal Antibody [clone: EPR22858-106] (Alexa Fluor® 555)
Catalog # ABCAAB318828-100UL
Supplier:  Abcam
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Seq:
    1286
  • Size:
    100 µl
  • Pack type:
    Vial

Specifications

About this item

Alexa Fluor® 555 Rabbit monoclonal [EPR22858-106] to RUNX2 - ChIP Grade.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: RUNX2
Clonality: Monoclonal
Clone: EPR22858-106
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: