Anti-CGRP-I+CGRP-II Rabbit Monoclonal Antibody [clone: EPR23804-95]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR23804-95] to CGRP-I+CGRP-II - BSA and Azide free.
Type: Primary
Antigen: CGRP-I+CGRP-II
Clonality: Monoclonal
Clone: EPR23804-95
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together