Anti-PD-L1 Rabbit Monoclonal Antibody [clone: 28-8] (Alexa Fluor® 555)
Catalog # ABCAAB213358-100
Supplier: Abcam
Specifications
- Size:100 µl
- Pack type:Vial
Specifications
About this item
Alexa Fluor® 555 Rabbit monoclonal [28-8] to PD-L1.
Type: Primary
Antigen: PD-L1
Clonality: Monoclonal
Clone: 28-8
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human