Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-IGHG4 Mouse Antibody [clone: MOPC-21] (Alexa Fluor® 647)
Anti-IGHG4 Mouse Antibody [clone: MOPC-21] (Alexa Fluor® 647)
Catalog # ABCAAB239459-100
Supplier:  Abcam
undefined
Anti-IGHG4 Mouse Antibody [clone: MOPC-21] (Alexa Fluor® 647)
Catalog # ABCAAB239459-100
Supplier:  Abcam

Specifications

  • Size:
    100 µg
  • Pack type:
    Vial

Specifications

About this item

Alexa Fluor® 647 Mouse IgG1 kappa [MOPC-21] - Isotype Control.

Type: Control
Antigen: IGHG4
Clonality:
Clone: MOPC-21
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: