About this item
Rabbit monoclonal [EPR24522-87] to CD64 - BSA and Azide free (Detector).
Type: Primary
Antigen: CD64
Clonality: Monoclonal
Clone: EPR24522-87
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- ABCAAB300685-100
- ABCAAB300685-1
- Antigen symbol:
- CD64
- CD64
- Antigen name:
- FCG1
- FCG1
- Antigen synonyms:
- FCGR1|IGFR1|FCGR1A|High affinity immunoglobulin gamma Fc receptor I|IgG Fc receptor I|Fc-gamma RI|Fc-gamma RIA|FcRI|FcgammaRIa
- FCGR1|IGFR1|FCGR1A|High affinity immunoglobulin gamma Fc receptor I|IgG Fc receptor I|Fc-gamma RI|Fc-gamma RIA|FcRI|FcgammaRIa
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR24522-87
- EPR24522-87
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular weight:
- 42632 Da
- 42632 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage temperature:
- Store at 4 °C.
- Store at 4 °C.
- Shipping temperature:
- 4 °C
- 4 °C
- Tested applications:
- sELISA
- sELISA