Human Recombinant GPRC5A (from HEK293 Cells)
New Product
Partial length Human GPRC5A, expressed in HEK293 cells.
: This product is for esearch use only. It is not for use in diagnostic or therapeutic procedures.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q8NFJ5
- Reconstitution instructions:Prior to reconstitution, centrifuge the vial at 5000 g for 3 - 5 minutes at room temperature. Reconstitute with appropriate volume of distilled sterile water to bring product to 1 mg/ml concentration. After addition of distilled sterile water, mix by gentle tapping. Note: It is not recommended to vortex or vigorously pipette the sample.
- Carrier-free:N
- Protein synonyms:G-protein coupled receptor family C group 5 member A|GPCR5A|PEIG-1|Phorbol ester induced gene 1|RAI3|RAIG-1|RAIG1|Retinoic acid-induced gene 1 protein|Retinoic acid-induced protein 3
- Protein/peptide name:GPRC5A
- Purity:>95% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 29,9 kDa after removal of the signal peptide The apparent molecular mass of GPRC5A-mFc is approximately 25 - 35 kDa due to glycosylation
- Sequence:GPRC5A (Met1-Thr33) + mFc (Pro99-Lys330)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE
Frequently Bought Together