Human Recombinant IL-16 (from E. coli)
Catalog # ANTIA317636-50
Supplier: Antibodies.com
New Product
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:1202
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# AAC12732.1
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:IL16|Pro-interleukin-16
- Protein/peptide name:IL-16
- Purity:>95% by SDS-PAGE under reducing conditions
- Molecular weight:13,4 kDa
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from a 0,2 µm filtered solution of 20 mM Phosphate Buffer, 150 mM NaCl, pH 7,0
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE
Specifications
About this item
Partial length Human IL-16, expressed in Escherichia coli.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.