

Bismuth lead tin cadmium alloy (50.0:26.7:13.3:10.0 w%), eutectic ≥99.9% (metals basis), ingot
Catalog # 40951.P2
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 lb
- Seq:7032
Specifications
About this item
:
Melting Pt:70 °C
:GHS08
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD00801077