


tert-Butyldimethylsilyl trifluoromethanesulphonate for synthesis, Sigma-Aldrich®
Catalog # 8.18313.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Molar Mass:264.34 g/mol
- Boiling Point:65 - 67 °C (13 hPa)
- Flash Point:36 °C
- Refractive Index:1.3848 (20 °C, 589 nm)
- Density:1.151 g/cm3 (20 °C)
Specification Test Results
About this item
:
:264.34 g/mol
ADR:8 (3),I
:M-Clarity™ quality level = MQ200
EINECS:274-102-0
UN:2920
:GHS02
:GHS05
:GHS07
Storage Temperature:Refrigerator
CAS Number:69739-34-0
Formula:C₇H₁₅F₃O₃SSi
MDL Number:MFCD00000405