Order Entry
Northern Ireland
ContactUsLinkComponent
Bismuth lead alloy (55.5:44.5 w%), eutectic ≥99.9% (metals basis), ingot
Health
Irritant
Bismuth lead alloy (55.5:44.5 w%), eutectic ≥99.9% (metals basis), ingot
Catalog # 40949.Q4
CAS Number:  
Health
Irritant
Bismuth lead alloy (55.5:44.5 w%), eutectic ≥99.9% (metals basis), ingot
Catalog # 40949.Q4
CAS Number:  

Specifications

  • Pk:
    0,25 lb
  • Seq:
    7020

Specifications

About this item

 :
Melting Pt:124 °C
 :GHS08
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD00801141