2-Methoxy-5-pyridineboronic acid 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (6-Methoxypyridin-3-yl)boronic acid
CAS Number:163105-89-3
:
Formula:C₆H₈BNO₃
MDL Number:MFCD02093044
Molecular Weight:152.95 g/mol
Melting Pt:132…133 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together