


o-Xylene, Hydranal™ solubilizer for Karl Fischer titration, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:95-47-6
:
Formula:C₈H₁₀
MDL Number:MFCD00008519
:106.17 g/mol
Merck Index:13,10136
Boiling Pt:143…145 °C (1,013 hPa)
Melting Pt:–25 °C
Density:0,88 g/cm³ (20 °C)
Flash Pt:7,6 %(V)
Storage Temperature:Room Temperature
EINECS:202-422-2
UN:1307
ADR:2,III
:GHS07
:GHS08
:GHS02
Specification Test Results
- Suitability (for KF-titration):test passed
- Assay of water:Max. 0.02 %
- Number Of Samples:Min. 2
Specification Test Results
Frequently Bought Together