Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide)
Catalog # MOLEM46709264
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:337.3 g/mol
EINECS:433-520-5
CAS Number:82911-69-1
Formula:C₁₉H₁₅NO₅
MDL Number:MFCD00010733