
1-Cyclopentene-1-carboxylic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1560-11-8
:
Formula:C₆H₈O₂
MDL Number:MFCD00001396
:112.13 g/mol
Boiling Pt:210 °C
Melting Pt:117…121 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Related Information
Frequently Bought Together