DSS (Disuccinimidyl suberate), Pierce™
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:68528-80-3
:
Formula:C₁₆H₂₀N₂O₈
MDL Number:MFCD00049059
:368.34 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Homobifunctional
- Reactive Moiety:NHS Ester
- Spacer:Mid-Length
- Form:Powder
- Quantity:10 x 2 mg/50 mg/1 g
- Type:Crosslinker
- Solubility:DMF, DMSO
- Spacer Arm:11.4 Å
Specification Test Results
Frequently Bought Together