Order Entry
Northern Ireland
ContactUsLinkComponent
β-Bromostyrene (mixture of cis and trans isomers) cis- and trans-mixture 97%
Irritant
β-Bromostyrene (mixture of cis and trans isomers) cis- and trans-mixture 97%
Certificates
undefined
Irritant
β-Bromostyrene (mixture of cis and trans isomers) cis- and trans-mixture 97%

Specification Test Results

  • Appearance:
    Clear yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6050 to 1.6090 (20°C, 589 nm)
  • GC:
    ≥96.0 % (sum of isomers)

Specification Test Results

Related Information

Frequently Bought Together