2,4-Dimethylpyrrole
Catalog # MOLE20588412-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:95.1424 g/mol
EINECS:210-912-2
CAS Number:625-82-1
Formula:C₆H₉N
MDL Number:MFCD00192088