2,4-Dimethylpyrrole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:625-82-1
:
Formula:C₆H₉N
MDL Number:MFCD00192088
:95.1424 g/mol
Boiling Pt:165…167 °C
Density:0,924 g/cm³
Flash Pt:>109 °C
EINECS:210-912-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow to orange Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4950 to 1.4990 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together