Order Entry
Northern Ireland
ContactUsLinkComponent
2,4,5-Triamino-6-hydroxypyrimidine sulphate for synthesis, Sigma-Aldrich®
2,4,5-Triamino-6-hydroxypyrimidine sulphate for synthesis, Sigma-Aldrich®
Catalog # 8.14764.0025
Supplier:  Merck
CAS Number:  
2,4,5-Triamino-6-hydroxypyrimidine sulphate for synthesis, Sigma-Aldrich®
Catalog # 8.14764.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    2 g/l (20 °C)
  • Melting Point:
    350 °C
  • Bulk Density:
    480 kg/m3
  • pH:
    2 - 3 (2 g/l, H2O, 20 °C)
  • Hill Formula:
    C4H9N5O5S
  • Chemical Formula:
    C4H7N5O . x H2SO4

Specification Test Results

About this item

 :
 :239.21 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:254-393-0
CAS Number:39267-74-8
Formula:C₄H₉N₅O₅S
MDL Number:MFCD00012785