ContactUsLinkComponent
Cut resistant sleeves, HyFlex® 72-265
Cut resistant sleeves, HyFlex® 72-265
Catalog # ANSE72-265
Supplier:  Ansell
undefined
Cut resistant sleeves, HyFlex® 72-265
Catalog # ANSE72-265
Supplier:  Ansell
Specifications
  • Garment application:
    Industrial
  • Risk area:
    Cuts
  • Colour:
    Grey
  • Length:
    380 mm
  • Seq:
    05
  • Fabric:
    Fiber glass/polyester/Dyneema®
  • Pk:
    1
Specifications
About this item
These sleeves contain a very high proportion of Dyneema® fibres which are as strong as steel fibres but much lighter and more flexible.
  • Sleeves provide optimal protection from cuts and abrasions
  • Light weight and comfortable fit ensure high levels of user acceptance
Ideal for use in plastics injection and moulding, paper processing and metal fabrication.
Certifications: EN 388:2016 (3X4XC), Cat. II