
Eriochrome cyanine R S. No. 840, Fluka™
Catalog # 32752-25G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Identity (IR):complying
- Suitability:complying
Specification Test Results
About this item
:
EINECS:222-641-7
:GHS07
Storage Temperature:Ambient
CAS Number:3564-18-9
Formula:C₂₃H₁₅Na₃O₉S
MDL Number:MFCD00012411