α-Naphtholphthalein, pure indicator
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 3,3-Bis(4-hydroxynaphthalen-1-yl)isobenzofuran-1(3H)-one
- CAS Number:596-01-0
- Formula:C₂₈H₁₈O₄
- MDL Number:MFCD00036202
- Molecular Weight:418.45 g/mol
- Melting Pt:240…250 °C
- Storage Temperature:Ambient
- EINECS:209-875-5
3 Options Available Below
Product Details & Documents
Synonyms: 3,3-Bis(4-hydroxynaphthalen-1-yl)isobenzofuran-1(3H)-one
- CAS Number:596-01-0
- Formula:C₂₈H₁₈O₄
- MDL Number:MFCD00036202
- Molecular Weight:418.45 g/mol
- Melting Pt:240…250 °C
- Storage Temperature:Ambient
- EINECS:209-875-5
Documents
ACRO19012
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Beige to pink to brown Powder
- Infrared spectrum:Conforms
- Loss on drying:≤5 % (110°C, 1 h)
- UV Lambda max:648 to 654 nm (in 0.1N NaOH)
- Specific absorbance A (1%/1cm):≥550 (at lamda max)
- Visual transition interval:from pH 7.3 (pink-yellow) to pH 8.7 (green-blue)
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



