
Menadione (2-methyl-1,4-naphthoquinone, vitamin K3) 98%
:58-27-5
:
:C₁₁H₈O₂
:MFCD00001681
:172.19 g/mol
:105…108 °C
:200-372-6
:GHS07
:Ambient
- Appearance:Yellow to green Powder
- Infrared spectrum:Conforms
- Melting point:105 to 108 °C
- HPLC:≥97.5 %
Frequently Bought Together