Fmoc-Met-Opfp
Catalog # 04-12-1512-5
Supplier: Novabiochem (Part of Merck)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:537.4983 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:86060-94-8
Formula:C₂₆H₂₀F₅NO₄S
MDL Number:MFCD00065665