



Benzylmagnesium chloride 1.4 M in THF, AcroSeal®
389551000.
:
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
- Appearance:Clear green-brown to brown-purple Solution (May develop some turbidity or precipitate)
- Titration Grignard:19 to 21 % (As BMC)
:
:150.89 g/mol
:–17 °C
:Ambient
:230-039-0
:2924
:3,II
:GHS07
:GHS05
:GHS02
:GHS08
:6921-34-2
:C₇H₇ClMg
:MFCD00000469