
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100165-88-6
:
Formula:C₄₈H₄₀P₂
MDL Number:MFCD01311709
:678.78 g/mol
Melting Pt:255…257 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together