
1-(2-Naphthoyl)-3,3,3-trifluoroacetone ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:893-33-4
:
Formula:C₁₄H₉F₃O₂
MDL Number:MFCD00054501
:266.22 g/mol
Melting Pt:74…76 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together