
N-Ethyldicyclohexylamine 96%
Catalog # ACRO314730050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4830 to 1.4870 (20°C, 589 nm)
- GC:≥95 %
Specification Test Results
About this item
:
Boiling Pt:114…115 °C (3 mmHg)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:230-534-1
UN:2735
ADR:8,III
:GHS05
CAS Number:7175-49-7
Formula:C₁₄H₂₇N
MDL Number:MFCD00003846