3-Nitrostyrene 97% stabilised
:586-39-0
:
:C₈H₇NO₂
:MFCD00007276
:149.1467 g/mol
:–5 °C
:1,07 g/cm³
:107 °C
:209-575-4
:GHS07
:Refrigerator
- Appearance:Clear yellow to yellow-green Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5820 to 1.5840 (20°C, 589 nm)
- GC:≥96.0 %
- Stabilizer:Approx. 0.1 % 3,5-di-tert-Butylcatechol
Frequently Bought Together