
Tetraethyl methylenebisphosphonate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1660-94-2
:
Formula:C₉H₂₂O₆P₂
MDL Number:MFCD00039887
:288.22 g/mol
Boiling Pt:171…174 °C (11 mmHg)
Density:1,16 g/cm³
Flash Pt:>110 °C
EINECS:216-764-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4400 to 1.4440 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together