

Ethyl dichlorophosphite 98%, rods
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1498-42-6
:
Formula:C₂H₅Cl₂OP
MDL Number:MFCD00000525
:146.94 g/mol
Boiling Pt:117…118 °C
Density:1,284
Flash Pt:4 °C (39 °F)
Storage Temperature:Refrigerator
EINECS:216-098-5
UN:2924
ADR:3,II
:GHS02
:GHS05
Related Information
Frequently Bought Together