
tert-Butyl (diethylphosphono)acetate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:27784-76-5
:
Formula:C₁₀H₂₁O₅P
MDL Number:MFCD00075414
:252.25 g/mol
Boiling Pt:100…103 °C (1,5 mmHg)
Density:1,074
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together