Order Entry
Northern Ireland
ContactUsLinkComponent
4,4'-Dibromooctafluorobiphenyl (DBOB) ≥99%
4,4'-Dibromooctafluorobiphenyl (DBOB) ≥99%
Catalog # B25126.03
CAS Number:  
4,4'-Dibromooctafluorobiphenyl (DBOB) ≥99%
Catalog # B25126.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:10386-84-2
 :
Formula:C₁₂Br₂F₈
MDL Number:MFCD00000310
 :455.94 g/mol
Melting Pt:113…115 °C
Storage Temperature:Ambient
EINECS:233-847-1
ADR:9,II