Order Entry
Northern Ireland
ContactUsLinkComponent
Neodymium(III) sulphate octahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Irritant
Neodymium(III) sulphate octahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Catalog # 13185.14
CAS Number:  
Irritant
Neodymium(III) sulphate octahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Catalog # 13185.14
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :720.78 g/mol (576.66 anhy)
Merck Index:14,6450
Melting Pt:1176 °C
Density:2,85
EINECS:233-262-1
 :GHS07
Storage Temperature:Ambient
CAS Number:13477-91-3
Formula:Nd₂S₃O₁₂·8H₂O
MDL Number:MFCD00149803