
trans-2,4-Dimethoxy-β-nitrostyrene ≥99%
Catalog # B20564.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:209.19 g/mol
Melting Pt:99…101 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1891-10-7
Formula:C₁₀H₁₁NO₄
MDL Number:MFCD00015922