


O-Dianisidine dihydrochloride suitable for use in glucose determination, Sigma-Aldrich®
Catalog # F5803-50MG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Appearance (Colour):White
- Appearance (Form):Crystals
- Solubility (Turbidity), 50 mg vial reconstituted with 20 ml of purified water:Clear
- Solubility (Colour), 50 mg vial reconstituted with 20 ml of purified water:Colourless
- Solubility (Form), 50 mg vial reconstituted with 20 ml of purified water:Solution
- pH:6.95 - 7.05
- Glucose Recovery, Recovery of 3 mg/ml NIST Glucose Standard:2.80 - 3.50 mg/ml
- Absorbance A450 Blank:≤0.05
- Completion Time:≤30 min
Specification Test Results
About this item
:
:317.21
Melting Pt:268 °C
UN:3261
ADR:8,III
:M-Clarity™ quality level = MQ200
EINECS:243-737-5
:GHS05
:GHS07
:GHS08
Storage Temperature:Refrigerator
CAS Number:20325-40-0
Formula:C₁₄H₁₈Cl₂N₂O₂
MDL Number:MFCD00012488