Order Entry
Northern Ireland
ContactUsLinkComponent
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester 95%
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester 95%
Catalog # H27036.03
CAS Number:  
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester 95%
Catalog # H27036.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :278.16 g/mol
Storage Temperature:Ambient
CAS Number:903550-26-5
Formula:C₁₄H₂₃BN₂O₃
MDL Number:MFCD09037501