Order Entry
Northern Ireland
ContactUsLinkComponent
(2R)-(-)-1-Benzoyl-2-tert-butyl-3-methyl-4-imidazolidinone for synthesis
(2R)-(-)-1-Benzoyl-2-tert-butyl-3-methyl-4-imidazolidinone for synthesis
Catalog # 8.14167.0001
Supplier:  Merck
CAS Number:  
(2R)-(-)-1-Benzoyl-2-tert-butyl-3-methyl-4-imidazolidinone for synthesis
Catalog # 8.14167.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    144 - 147 °C
  • Molar Mass:
    260.33 g/mol

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
CAS Number:101055-57-6
Formula:C₁₅H₂₀N₂O₂