



Borane tetrahydrofuran complex 1 M in THF stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14044-65-6
:
Formula:C₄H₁₁BO
MDL Number:MFCD00012429
:85.94 g/mol
Density:0,876 g/cm³
Flash Pt:–22 °C
Storage Temperature:Refrigerator
EINECS:237-881-8
UN:3399
ADR:4.3,I
:GHS02
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear colorless Solution (May develop some turbidity or precipitate)
- Stabilizer:≤0.005 M sodium borohydride
- Strength:0.95 to 1.10 M
- Density:1.690 to 1.730 kg/l
- Viscosity:0.39 to 0.49 cSt (at 25°C)
- Dielectric strength:≥35 kV
- Distillation range:≥95 % (between 47-57°C)
- Pour point:< -60°C
- Residue:≤1 ppm
Specification Test Results
Related Information
Frequently Bought Together