2-Hydroxypyridine N-oxide
Catalog # MOLEM39979413
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:111.10 g/mol
EINECS:236-100-8
CAS Number:13161-30-3
Formula:C₅H₅NO₂
MDL Number:MFCD00006195