Order Entry
Northern Ireland
ContactUsLinkComponent
O-Ethylhydroxylamine hydrochloride
O-Ethylhydroxylamine hydrochloride
Catalog # MOLEM21172868
Supplier:  Molekula
CAS Number:  
O-Ethylhydroxylamine hydrochloride
Catalog # MOLEM21172868
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Ethoxyamine hydrochloride
 :
 :97.54 g/mol
EINECS:222-060-9
CAS Number:3332-29-4
Formula:C₂H₈ClNO
MDL Number:MFCD00012956