Order Entry
Northern Ireland
ContactUsLinkComponent
Ethyl-N-hydroxyacetimidate 97%
Irritant
Ethyl-N-hydroxyacetimidate 97%
Certificates
undefined
Irritant
Ethyl-N-hydroxyacetimidate 97%

Specification Test Results

  • Appearance:
    Colorless to yellow Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4320 to 1.4350 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together