

n-Butyl isocyanide 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2769-64-4
:
Formula:C₅H₉N
MDL Number:MFCD00000008
:83.132 g/mol
Density:0,79 g/cm³
Storage Temperature:Refrigerator
EINECS:220-458-7
UN:1935
ADR:6.1,II
:GHS06
:GHS08
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3950 to 1.3970 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together